Learn More
MilliporeSigma™ Supelco™ Static Dilution Bottle with Mininert™ Valve
Catalog No. 111004417
Change view
Click to view available options
Quantity:
1/Pk.
Catalog No. Quantity
111004417 1/Pk.
1 options
Catalog No. 111004417 Supplier MilliporeSigma™ Supelco™ Supplier No. 21992
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Static dilution bottle with Mininert™™ valve features a two liter round bottomed flask with a threaded neck. Use the static dilution bottle to prepare gaseous volatile organic standards, using a technique developed by the US EPA for air analyses.
Specifications
Quantity 1/Pk.
Mininert is a registered trademark of Valco Instruments Co. Inc.