missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STAT2 Monoclonal Antibody (K24015_11D7), Invitrogen™

Catalog No. ma556198
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556198 200 μL
MA555818 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556198 Supplier Invitrogen™ Supplier No. MA556198
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: DEAFGCYYQE KVNLQERRKY LKHRLIVVSN RQVDELQQPL ELKPEPELES LELELGLVPE PELSLDLEPL LKAGLDLGPE LESVLESTLE PVIEPTLCMV

STAT2 is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. In response to interferon (IFN), this protein forms a complex with STAT1 and IFN regulatory factor family protein p48 (ISGF3G), in which this protein acts as a transactivator, but lacks the ability to bind DNA directly. Transcription adaptor P300/CBP (EP300/CREBBP) has been shown to interact specifically with this protein, which is thought to be involved in the process of blocking IFN-alpha response by adenovirus.

Specifications

Antigen STAT2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K24015_11D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Stat2
Gene Accession No. P52630
Gene Alias 1110034C02Rik; 1600010G07Rik; acute phase response factor; acute-phase response factor; ADMIO; ADMIO1; Aprf; AW109958; AW496480; DNA-binding protein APRF; EGK_03803; HIES; IMD44; interferon alpha induced transcriptional activator; ISGF-3; MGC59816; p113; signal transducer and activator of transcription 2; signal transducer and activator of transcription 2, 113kDa; signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor); STAT; STAT 2; STAT113; STAT2; STAT-2; Stat3
Gene Symbols Stat2
Host Species Mouse
Immunogen Recombinant human STAT2 (immunogen range is 671-770).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6773
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less