missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Starter pH Pen Meters
Convenient pH testing. OHAUS Starter pH Pen Meters offer a convenient way to measure pH, or the relative acidity or alkalinity of a sample at a given temperature. The pH pen meters can be completely operated with just one hand, freeing up your other hand to handle samples.
$258.21 - $267.87
Specifications
| Accuracy (pH) | ±0.1 |
|---|---|
| Includes | 4 1.5V Batteries |
| For Use With (Application) | Measuring pH, or the relative acidity or alkalinity of a sample at a given temperature |
| Calibration | Automatic/Manual |
| Range (pH) | 0 to 14 |
Description
- Fast, straightforward results
- Durable IP67 waterproof design
- Equipped with wrist strap for security
- Automatic Shutdown after six minutes of non-use
- Requires four AG13 1.5V Micro Alkaline Batteries
- Size/Weight: 185 x 45 x 38 mm, 110 g
- Includes buffer powder pack (ST20)
Specifications
| ±0.1 | |
| Measuring pH, or the relative acidity or alkalinity of a sample at a given temperature | |
| 0 to 14 | |
| None | |
| Automatic Data Logging | |
| 0.24 lb. | |
| Portable pH Meter |
| 4 1.5V Batteries | |
| Automatic/Manual | |
| 3 | |
| 30 | |
| 110 g | |
| 4 x AG13 1.5 V Micro Alkaline |