missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Starter Pak No. 2
GREENER_CHOICE

Catalog No. 215367
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
215367 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 215367 Supplier Gibco™ Supplier No. 215367

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contain a variety of animal origin and animal origin-free peptones that offer many essential nutrients needed for the production of human and animal vaccines.

  • Provides conveniently packaged, 100-g, non-GMP samples of these peptones that are best suited for vaccine production and contains:
    • Bacto Yeast Extract—this yeast has the highest level of carbohydrates of our yeast products and works well in a variety of human and animal health vaccines
    • Phytone Peptone and Difco Soytone—both enzymatic digests of soy, these peptones also are a nutritious source of carbohydrates. These products work well in microbial fermentation processes as well as in mammalian cell culture processes such as CHO. Blending these soy peptones with yeast peptones has been shown to provide additional benefit to cultures.
    • Bacto Proteose Peptone No. 2 and Bacto Proteose Peptone No 3—these enzymatic digests of porcine protein provide nutrition for fastidious microorganisms. Bacto Proteose Peptone No. 3 can replace serum in many applications and helps increase monoclonal antibody and recombinant protein production in CHO cells.
    • Bacto Casamino Acids—this supplement has low salt and iron content, making it an excellent supplement for media formulations for which nitrogen requirements are minimal

Specifications

Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Human and animal vaccines
Quantity 1 Ea.
Source Animal origin and animal origin-free