Learn More
Eisco™ Starter Kit for Simple Organic Chemistry - 8 Pieces
The ideal set for organic chemistry labs, distillation, fractional distillation, separation, purification, and synthesis. This Starter kit for Simple Organic Chemistry comprises 8 items of 14/23 ground joint. All pieces made with high quality borosilicate glass.
Supplier:  Eisco™ CH0881
Catalog No. S28014
Only null left
Description
Description
  • The ideal starter set for organic chemistry labs, distillation, fractional distillation, separation, purification, and synthesis.
  • All pieces made of 3.3 borosilicate glass that is hand blown and polished.
  • Metal ware stands not included in set.
  • Recommended stands are Eisco: MTST1.
  • This set comprises 8 items of 14/23 ground joint: Beaker 25 mL, Vigreux Column 100 mm, Liebig Condenser 100 mm, Still head Screw cap, Adapter, Receiver Adapter, Pear shaped Flask 25 mL, Thermometer -10°C to +110°C.
Specifications
Specifications
Borosilicate Glass
14/23
Chemistry Kit
8
Non-sterile
Microchemistry Kits
Product Suggestions
Product Suggestions