missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Starsys 34 in. Tall Undercounter Storage Cart
Undercounter storage carts enhance productivity and space efficiency.
Supplier: Metro™ SXR34NL2
Description
- Durable easy-to- clean, polymer enclosures will not chip, peel, or rust.
- Robust and stable structure accommodates even the most sensitive equipment.
- Security options including non-locking, key locking, and electronic PIN locking with key override.
Chemical Identifiers
| Not Autoclavable | |
| 594.36 mm | |
| Polymer | |
| 548.64 mm |
| Taupe | |
| 864.87 mm | |
| Product will be free from defects in material and workmanship under normal use, service and maintenance for a period of one (1) year from the date of shipment of the Product by Seller, unless otherwise specified by Seller in writing. |
Specifications
| Not Autoclavable | |
| Taupe | |
| 864.87 mm | |
| Product will be free from defects in material and workmanship under normal use, service and maintenance for a period of one (1) year from the date of shipment of the Product by Seller, unless otherwise specified by Seller in writing. | |
| 548.64 mm |
| Cart with Door | |
| 594.36 mm | |
| Polymer | |
| 95.2 lb. |