missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Starsys 34 in. Tall Undercounter Storage Cart

Undercounter storage carts enhance productivity and space efficiency.

Supplier:  Metro™ SXR34KL2

 View more versions of this product

Catalog No. 13460295

Only null left

Description

Description

  • Durable easy-to- clean, polymer enclosures will not chip, peel, or rust.
  • Robust and stable structure accommodates even the most sensitive equipment.
  • Security options including non-locking, key locking, and electronic PIN locking with key override.
Specifications

Chemical Identifiers

Not Autoclavable
594.36 mm
Polymer
548.64 mm
Taupe
864.87 mm
Product will be free from defects in material and workmanship under normal use, service and maintenance for a period of one (1) year from the date of shipment of the Product by Seller, unless otherwise specified by Seller in writing.

Specifications

Not Autoclavable
Taupe
864.87 mm
Product will be free from defects in material and workmanship under normal use, service and maintenance for a period of one (1) year from the date of shipment of the Product by Seller, unless otherwise specified by Seller in writing.
548.64 mm
Cart with Door
594.36 mm
Polymer
87.9 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions