missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch, Soluble, Powder, Reagent, ACS, Spectrum™ Chemical
S1555, 9005-84-9, (C6H10O5)n
Supplier: Spectrum Chemical Mfg Cor S155512KG
Description
Spectrum™ Chemical Starch, Soluble, Powder, Reagent, ACS is indicator for redox titrations where triiodide is present. Starch forms a very dark blue-black complex with triiodide which can be made by mixing iodine with iodide (often from potassium iodide). The color of the starch complex is so deep, that it can be detected visually when the concentration of the iodine is as low as 0.00002 M at 20 °C. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Specifications
| 9005-84-9 | |
| 5.0 to 7.0 | |
| Poly Pail | |
| 12 kg | |
| OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O | |
| 342.30 |
| 1 | |
| 0.004 | |
| C12H22O11 | |
| GUBGYTABKSRVRQ-ASMJPISFSA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol | |
| ACS |
Chemical Identifiers
| 9005-84-9 | |
| 342.30 | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol |
| C12H22O11 | |
| GUBGYTABKSRVRQ-ASMJPISFSA-N | |
| OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O |