missing translation for 'onlineSavingsMsg'
Learn More

Starch, Soluble, Powder, Reagent, ACS, Spectrum™ Chemical

Catalog No. p-7158314
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607003 2.5 kg Poly Pail
18607004 12 kg Poly Pail
18607005 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18607003 Supplier Spectrum Chemical Mfg Cor Supplier No. S15552.5KG
Only null left

S1555, 9005-84-9, (C6H10O5)n

Spectrum™ Chemical Starch, Soluble, Powder, Reagent, ACS is indicator for redox titrations where triiodide is present. Starch forms a very dark blue-black complex with triiodide which can be made by mixing iodine with iodide (often from potassium iodide). The color of the starch complex is so deep, that it can be detected visually when the concentration of the iodine is as low as 0.00002 M at 20 °C. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Identifiants chimiques

CAS 9005-84-9
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
InChI Key GUBGYTABKSRVRQ-ASMJPISFSA-N
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

Spécifications

CAS Max % 1
pH 5.0 to 7.0
Ignition Residue 0.004
Packaging Poly Pail
Quantity 2.5 kg
Grade ACS