missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch Indicator, 2% (w/v), Mercury Free, for Iodometric Titrations, Ricca Chemical
Iodometric Titrations
$271.18 - $2922.70
Specifications
| Chemical Name or Material | Starch Indicator |
|---|---|
| CAS | 7732-18-5 , 9005-84-9 , 69-72-7 |
| CAS Min % | 96.0,1.92,0.19 |
| CAS Max % | 100.0,2.0,0.2 |
| Concentration | 2% (w/v) |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
8055-16
|
Ricca Chemical Company
805516* |
500 mL | Natural Poly Bottle |
Each for $271.18
|
|
||||
|
805532
|
Ricca Chemical Company
805532 |
1 L | Natural Poly Bottle |
Each for $403.07
|
|
||||
|
8055-1
|
Ricca Chemical Company
R80550004A |
4 L | Natural Poly Bottle |
Each for $983.90
|
|
||||
|
8055-5
|
Ricca Chemical Company
80555 |
20 L | Cubitainer |
Each for $2,922.70
|
|
||||