missing translation for 'onlineSavingsMsg'
Learn More

Starch Indicator, 1% (w/v), Mercury Free, for Iodometric Titrations, Ricca Chemical

Iodometric Titrations

Supplier:  Ricca Chemical Company R80500004F

 View more versions of this product

Catalog No. R80500004F


Only null left

Description

Description

Specifications

Specifications

Starch Indicator
97.0,0.97,0.12
1% (w/v)
Indicator
Aqueous Solution
5 to 7
Liquid
7732-18-5 , 9005-84-9 , 69-72-7
100.0,1.01,0.13
1% W/V
Cubitainer
White
4 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions