missing translation for 'onlineSavingsMsg'
Learn More

Starch Indicator, 1% (w/v) Aqueous Solution, Stabilized, Spectrum™ Chemical

Catalog No. 18614064
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614064 20 L Poly Bottle
18614061 500 mL Poly Bottle
18614062 1 L Poly Bottle
18614063 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614064 Supplier Spectrum Chemical Mfg Cor Supplier No. S57220LT
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 9005-25-8 , 7732-18-5 , 9005-84-9

Specifications

Concentration 1%
Grade Chemical Solution
Packaging Poly Bottle
Color Colorless to White
Quantity 20 L
Chemical Name or Material Starch Indicator

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.