missing translation for 'onlineSavingsMsg'
Learn More

Starch Indicator, 1% (w/v) Aqueous Solution, Stabilized, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S5721LT

 View more versions of this product

Catalog No. 18614062

Only null left

Description

Description

Specifications

Specifications

Starch Indicator
1%
Poly Bottle
1 L
9005-25-8 , 7732-18-5 , 9005-84-9
Chemical Solution
Colorless to White

Chemical Identifiers

9005-25-8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.