accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Biochemical Reagents
Staining Reagents
Stains and Dyes
R7999200120
Print
Starch Indicator, 0.2% (w/v), Ricca Chemical
Supplier:
Ricca Chemical Company
R7999200120A
Catalog No.
R7999200120
Sign In
or
Register
to check your price and availability.
$80.43
/
Each
Quantity
Only null left
Add to cart
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Promotions
Description
Description
Specifications
Specifications
Chemical Name or Material
Starch Indicator
CAS Min %
98.0,0.2,0.1
Concentration
0.2% (w/v)
Grade
Indicator
Solvent or Matrix
Aqueous Solution
pH
5 to 7
Physical Form
Liquid
CAS
7732-18-5
,
9005-84-9
,
69-72-7
CAS Max %
100.0,0.2,0.1
Percent Purity
0.2% W/V
Packaging
Natural Poly Bottle
Color
White
Quantity
120 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions