missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stannous Chloride, Dihydrate, Crystal, Reagent, ACS, 98-103%, Spectrum™ Chemical
S1535, 10025-69-1, SnCl2·2H2O
$248.50 - $18256.57
Chemical Identifiers
| CAS | 10025-69-1 |
|---|---|
| Molecular Formula | Cl2H4O2Sn |
| Molecular Weight (g/mol) | 225.64 |
| InChI Key | FWPIDFUJEMBDLS-UHFFFAOYSA-L |
| IUPAC Name | λ²-tin(2+) dihydrate dichloride |
| SMILES | O.O.[Cl-].[Cl-].[Sn++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606985
|
Spectrum Chemical Mfg Cor
S153525GM |
25 g | Poly Bottle |
Each for $248.50
|
|
||||
|
18606986
|
Spectrum Chemical Mfg Cor
S1535125GM |
125 g | Poly Bottle |
Each for $522.80
|
|
||||
|
18606988
|
Spectrum Chemical Mfg Cor
S1535500GM |
500 g | Poly Bottle |
Each for $1,130.83
|
|
||||
|
18606987
|
Spectrum Chemical Mfg Cor
S1535125GMCS6 |
6 x 125 g | Poly Bottle |
Case of 6 Each for $2,170.37
|
|
||||
|
18606989
|
Spectrum Chemical Mfg Cor
S1535500GMCS6 |
6 x 500 g | Poly Bottle |
Case of 6 Each for $5,571.47
|
|
||||
|
18606991
|
Spectrum Chemical Mfg Cor
S153525KGCS4 |
4 x 2.5 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18606992
|
Spectrum Chemical Mfg Cor
S153512KG |
12 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18606993
|
Spectrum Chemical Mfg Cor
S153550KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Stannous Chloride, Dihydrate, Crystal, Reagent, ACS is also known as Tin Chloride, has a white solid crystal like aspect. Mostly used in electrolytic baths destined for tin-plating, it is also widely used as a reducing agent. In the textile industry it is employed as a mordant to increase the brightness of certain colors in the fabric. It is also employed by metal enthusiasts to determine platinum and gold group metals. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.Chemical Identifiers
| 10025-69-1 | |
| 225.64 | |
| λ²-tin(2+) dihydrate dichloride |
| Cl2H4O2Sn | |
| FWPIDFUJEMBDLS-UHFFFAOYSA-L | |
| O.O.[Cl-].[Cl-].[Sn++] |
Specifications
| 10025-69-1 | |
| 25 g | |
| FWPIDFUJEMBDLS-UHFFFAOYSA-L | |
| λ²-tin(2+) dihydrate dichloride | |
| 98 to 103% | |
| Poly Bottle |
| 1 | |
| Cl2H4O2Sn | |
| O.O.[Cl-].[Cl-].[Sn++] | |
| 225.64 | |
| ACS |