missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride, Dihydrate, Crystal, Reagent, ACS, 98-103%, Spectrum™ Chemical

Catalog No. 18606993
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Poly Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606993 50 kg Poly Bottle
18606985 25 g Poly Bottle
18606986 125 g Poly Bottle
18606988 500 g Poly Bottle
18606987 6 x 125 g Poly Bottle
18606989 6 x 500 g Poly Bottle
18606991 4 x 2.5 kg Poly Bottle
18606992 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18606993 Supplier Spectrum Chemical Mfg Cor Supplier No. S153550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1535, 10025-69-1, SnCl2·2H2O

Spectrum™ Chemical Stannous Chloride, Dihydrate, Crystal, Reagent, ACS is also known as Tin Chloride, has a white solid crystal like aspect. Mostly used in electrolytic baths destined for tin-plating, it is also widely used as a reducing agent. In the textile industry it is employed as a mordant to increase the brightness of certain colors in the fabric. It is also employed by metal enthusiasts to determine platinum and gold group metals. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2H4O2Sn
Molecular Weight (g/mol) 225.64
InChI Key FWPIDFUJEMBDLS-UHFFFAOYSA-L
IUPAC Name λ2-tin(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Sn++]

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 50 kg
Percent Purity 98 to 103%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Stannous Chloride, Dihydrate, Crystal