Learn More
Fisherbrand™ Standing Height Wood Corner Filler, Adjustable
Fisherbrand™ Wood Corner Fillers bring quality, warmth and durability to your laboratory.
 View more versions of this product
Catalog No. FB9135T12M7
Only null left
Explore available promotions
Description
Description
  • Made from hand selected premium rotary cut maple or rift cut oak veneers
  • Robotically applied environmentally friendly finishes are chemically resistant.
  • Corner Fillers are used to close the gap between two cabinets in a corner.
  • Fillers can be cut to fit in the field
Compliance
SEFA 8
Specifications
Specifications
Mocha Stain
2 to 12 in.
35 in.
2 to 12 in.
Wood (Maple Veneer)
5.1 to 30.5 cm
89.2 cm
5.1 to 30.5 cm
Product Suggestions
Product Suggestions