Learn More
Fisherbrand™ Standing Height Rear and Center Wood Fillers
Fisherbrand™ Rear and Center Fillers bring quality, warmth and durability to your laboratory.
 View more versions of this product
Catalog No. FB903518M7
Only null left
Explore available promotions
Description
Description
  • Made from hand selected premium rotary cut maple or rift cut oak veneers
  • Robotically applied environmentally friendly finishes are chemically resistant.
  • Rear and Center Fillers are used between back-to-back base cabinets or to close space at the rear of a cabinet.
  • Fillers can be cut to fit in the field
Compliance
SEFA 8
Specifications
Specifications
Mocha Stain
1 in.
35 in.
2 to 18 in.
Wood (Maple Veneer)
2.5 cm
88.9 cm
5.1 to 45.7 cm
Product Suggestions
Product Suggestions