missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Standardization Pipet Tips
Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip
Description
Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip
- Soft pliable tip collar allows this tip to fit on pipets made by various manufacturers
- Lower cost and increased convenience by purchasing fewer products
- Available in a variety of packaging options
- Eliminate the need to buy pipetter-specific tips
- Certified by independent laboratory
- Nonsterile: RNase, DNase, DNA and pyrogen free
Chemical Identifiers
| 0.1 to 200 μL | |
| 5cm | |
| Universal |
| Autoclavable | |
| DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free | |
| Polypropylene |
Specifications
| 0.1 to 200 μL | |
| Yellow | |
| 2 in. | |
| DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free | |
| Universal | |
| Universal Fit |
| Non-sterile | |
| Autoclavable | |
| 5cm | |
| Rack | |
| Lift-off Rack, 10 x 96 | |
| Polypropylene |