missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Standardization Pipet Tips
GREENER_CHOICE

Catalog No. p-4906597
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Natural
Yellow
Packaging:
Bulk
Bulk Pack
Lift-off rack, 10 x 96
Stack Rack, 5 x 200
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Color Packaging
02681134 Non-sterile Natural Bulk
02681135 Non-sterile Natural Lift-off rack, 10 x 96
02681141 Non-sterile Yellow Lift-off rack, 10 x 96
02681140 Non-sterile Yellow Bulk Pack
02681142 Sterile Yellow Lift-off rack, 10 x 96
213752 Sterile Natural Stack Rack, 5 x 200
2119727 Sterile Yellow Stack Rack, 5 x 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 02681134
Only null left
Edge

Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip

  • Soft pliable tip collar allows this tip to fit on pipets made by various manufacturers
  • Lower cost and increased convenience by purchasing fewer products
  • Available in a variety of packaging options
  • Eliminate the need to buy pipetter-specific tips
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA and pyrogen free

Packaging Code -- BP: Bulk Pack; RI: Rack Inserts; HR: Hinged Rack; HRS: Hinged Rack; HR/T: Hinged Rack, Trace Metal-free; LR: Lift-off Rack; LRS: Lift-off Rack; SR: Stack Rack; IWS: Individually Wrapped.

Specifications

Volume (Metric) 0.1 to 200 μL
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Length (English) 2 in.
Length (Metric) 5cm
Clean Claims DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free
Format Bulk
Tip Style Universal
Packaging Bulk
For Use With (Equipment) Universal Fit
Material Polypropylene
Show More Show Less