missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Standard Water (Fresh Water Substitute), Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Standard Water (Fresh Water Substitute) |
| CAS | 7732-18-5, 144-55-8, 7791-18-6, 10035-04-8, 7732-18-5 |
| Color | Colorless |
| Grade | Laboratory |
| Packaging | Cubitainer |
| Quantity | 10 L |
| Physical Form | Liquid |