Learn More
Thermo Scientific Chemicals Standard Sample Tube, 3.24mm ID
Catalog No. AA14349KL
Change view
Click to view available options
Quantity:
20 Pack
5 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA14349KL 20 Pack
AA14349KJ 5 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA14349KL Supplier Thermo Scientific Chemicals Supplier No. 014349KL
For use with MKII Magnetic Susceptibility Balances
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • I.D.: 3.24mm
Specifications
Quantity 20 Pack
Type Standard Sample Tube
RUO – Research Use Only