missing translation for 'onlineSavingsMsg'
Learn More

Standard mOsm Solutions, Precision Systems

Catalog No. 23268952
Change view
Click to view available options
Concentration or Composition Notes:
100 mOsm/kg H2O
1000 mOsm/kg H2O
1100 mOsm/kg H2O
1500 mOsm/kg H2O
300 mOsm/kg H2O
500 mOsm/kg H2O
900 mOsm/kg H2O
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration or Composition Notes
23268952 1000 mOsm/kg H2O
1282712 100 mOsm/kg H2O
1282713 300 mOsm/kg H2O
1282714 500 mOsm/kg H2O
1282716 900 mOsm/kg H2O
1282718 1100 mOsm/kg H2O
1282720 1500 mOsm/kg H2O
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 23268952 Supplier Precision Systems Inc Supplier No. PSY2110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Osmometry Standard
Concentration or Composition Notes 1000 mOsm/kg H2O
Physical Form Liquid
Quantity 125 mL