missing translation for 'onlineSavingsMsg'
Learn More

Standard Mixture of n-Paraffins (consists of C12, C13, C14, C15 and C16) [Standard Material for GC], TCI America™

Catalog No. S04555ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04555ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04555ML Supplier TCI America Supplier No. S04555ML
Only null left

Chemical Identifiers

MDL Number MFCD00197832
Synonym n-Paraffin Mixture

Specifications

Physical Form Liquid
Color Colorless
Quantity 5 mL
Chemical Name or Material Standard Mixture of n-Paraffins (consists of C12, C13, C14, C15 and C16) [Standard Material for GC]