missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Standard Mixture of n-Paraffins (consists of C10, C11, C12, C13 and C14) [Standard Material for GC], TCI America™
Specifications
Specifications
| Physical Form | Liquid |
| Color | Colorless |
| Quantity | 5 mL |
| Chemical Name or Material | Standard Mixture of n-Paraffins (consists of C10, C11, C12, C13 and C14) [Standard Material for GC] |
Safety and Handling
| TSCA | No |