missing translation for 'onlineSavingsMsg'
Learn More

Standard Mixture of Fatty Acid Methyl Esters, TCI America™

Catalog No. S04595ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04595ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04595ML Supplier TCI America Supplier No. S04595ML
Only null left

(consists of Methyl Hexanoate, Heptanoate, Octanoate, Nonanoate, and Decanoate), [Standard Material for GC]

Chemical Identifiers

MDL Number MFCD00197826
Synonym Fatty Acid Methyl Ester Mixture

Specifications

Quantity 5 mL
Chemical Name or Material Standard Mixture of Fatty Acid Methyl Esters (consists of Methyl Hexanoate, Heptanoate, Octanoate, N