missing translation for 'onlineSavingsMsg'
Learn More

American Biotech Supply Standard Manual Defrost Laboratory Freezer with Natural Refrigerants
GREENER_CHOICE

Manual defrost freezers offer a cold-storage solution for a variety of laboratory needs. Units feature a series of temperature alarms to keep stored products at peak effectiveness. These freezers come equipped with keyed door locks, interior shelving, probe access, and microprocessor temperature controllers.

$6,613.11 - $12,569.21

Specifications

Type Laboratory Freezer
Temperature Range -15°C to -25°C
Temperature Stability ±0.1°C
Door Style Solid Swing
No. of Doors 1
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (English) Amperage Min. Clearance Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Amperage Min. Clearance Price Quantity & Availability  
22651377
GREENER_CHOICE
View Documents Promotion Details
American Biotech Supply
ABTHCMFP14
14 cu. ft. 3.5 A 74 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22651378
GREENER_CHOICE
View Documents Promotion Details
American Biotech Supply
ABTHCMFP17
17 cu. ft. 6 A 68 in.
Each for $6,613.11
Only null left
 
Description

Description

Standard Manual Defrost Laboratory Freezers with Natural Refrigerants offer a cold-storage solution for a variety of laboratory needs. Units feature a series of temperature alarms to keep stored products at peak effectiveness and a two year parts and labor warranty, plus an additional three year compressor parts warranty.

  • Microprocessor Temperature Controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Remote alarm contacts
  • Interior shelving
  • Probe access port (3/8 in.)
  • 2 probes; 1 in air and 1 in glycol bottle
  • Keyed door lock
  • High density urethane foam cabinet and door insulation
  • Powder coat exterior
  • Natural, hydrocarbon refrigerant
Specifications

Specifications

Laboratory Freezer
±0.1°C
1
Scratch-resistant Painted Steel
Plastic
6 Fixed
Upright
White
Two Year Parts and Labor Warranty, Plus Additional Three Year Compressor Parts Warranty
Audible and Visual High and Low Temperature Alarms
115 V
Natural, Hydrocarbon
-15°C to -25°C
Solid Swing
Right
Polyurethane Foam
Powder-coated Steel
cULus
Digital Temperature Display
Fan-forced
Manual
NEMA 5-15 P
Bottom-mounted
Videos
SDS
Documents

Documents