Learn More
METTLER TOLEDO Standard MA-P Precision Balance
Catalog No. 01804218
Change view
Click to view available options
Capacity:
620 g
2200 g
6.2 kg
Readability:
10 mg
0.1 g
Model:
MA2002P
MA6001P
MA602P
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Readability Model
01804218 620 g 10 mg MA602P
01804216 2200 g 10 mg MA2002P
01804214 6.2 kg 0.1 g MA6001P
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01804218 Supplier METTLER TOLEDO Supplier No. 30697487
Earn Up to 5X the Fisher Scientific Buck$ with Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
METTLER TOLEDO Standard MA-P Precision Balances are simple to operate and portable for on-the-go weighing, without compromising on accuracy or performance.
The precise weigh cell delivers highly accurate results with each use. Increase efficiency in your workflow with the intuitive touchscreen that guides you through each weighing procedure. Easily export data to a connected printer, computer, or USB device.
MA-P Balances include:
  • Strain gauge weighing cell
  • Internal adjustment
  • LCD hybrid touchscreen
  • 10 built-in applications
  • Power cable and battery operated
  • Front level indicator
  • Overload protection
  • 2 interfaces: USB-A, RS232
  • Bluetooth option
The MA Compact is the successor to the PL-E Balance. See how it ranks among our balance lines:
  • XPR Excellence Precision Balances
  • MX Advanced Precision Balances
  • MR Advanced Precision Balances
  • MA Standard Precision Balances
  • ME Standard Precision Balances
  • LA Standard Precision Balances
Specifications
Connection Type USB-A, RS232
Capacity 620 g
Display Type Touchscreen
Readability 10 mg
Material PBT
Model MA602P
Stabilization Time 1.5 sec.
Type Precision Balance
Electrical Requirements 100/240 V, 50/60 Hz, or 8 AA Batteries
Legal for Trade No