Learn More
METTLER TOLEDO Standard MA Analytical Balance
METTLER TOLEDO Standard MA Analytical Balances are highly accurate, efficient, simple to operate and designed with robust construction for proven long lasting performance.
Supplier:  METTLER TOLEDO 30697411
 View more versions of this product
Catalog No. 01-804-250
Earn Up to $2,000 in Accessories with Purchase
View all promotions
Only null left
Description
Description
The LCD hybrid touchscreen is simple to operate and will increase efficiency of your daily tasks with 9 built-in applications and statistical data analysis. Built with a metal base, PBT top housing and overload protection to defend against rough handling in any weighing environment, ensuring quality performance for years to come.
MA Analytical Balances include:
  • Internal adjustment
  • LCD hybrid touchscreen with onscreen instructions
  • 9 built-in applications
  • Statistical data analysis
  • Front level indicator
  • Metal base, PBT top housing
  • Overload protection
  • 2 interfaces: USB-A, RS232
  • Bluetooth option
The MA Balance is the successor to the ME-T Balance. See how it ranks among our balance lines:
  • XPR Excellence Analytical Balances
  • MX Advanced Analytical Balances
  • MR Advanced Analytical Balances
  • MA Standard Analytical Balances
  • ME Standard Analytical Balances
  • LA Standard Analytical Balances
Specifications
Specifications
USB-A, RS232
Touchscreen
Metal, PBT
2 sec.
100/240 V, 50/60 Hz
220 g
0.1 mg
MA204
Analytical Balance
No
Product Suggestions
Product Suggestions