missing translation for 'onlineSavingsMsg'
Learn More

Standard Hard Water, 200 ppm Hardness as CaCO3, Ricca Chemical

Supplier:  Ricca Chemical Company R348420020F

Catalog No. R348420020F


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hard Water Standard
89-83-8
Liquid
Colorless
100°C
Infinitely soluble in water
Analytical Standard
CC1=CC(=C(C=C1)C(C)C)O
150.221
6989
Laboratory
Cubitainer
Thymol odor
C10H14O
0.0°C
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Calibration Standard
MGSRCZKZVOBKFT-UHFFFAOYSA-N
5-methyl-2-propan-2-ylphenol
20 L
CHEBI:27607

Chemical Identifiers

89-83-8
150.221
6989
5-methyl-2-propan-2-ylphenol
C10H14O
MGSRCZKZVOBKFT-UHFFFAOYSA-N
CHEBI:27607
CC1=CC(=C(C=C1)C(C)C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : General

SDS
Documents

Documents

Promotions

Promotions