Learn More
Corning™ Standard Conical Bioprinting Nozzles
These Standard Conical Bioprinting Nozzles are designed fo use with Matribot™ Bioprinter. They are made from polypropylene, have a female Luer connector, and are available in 22G, 25G, and 27G.
Supplier:  Corning™ 6165
 View more versions of this product
Catalog No. 01257115
Only null left
Description
Description
  • Made of polypropylene
  • Feature a female Luer connector
  • Available in 22 G, 25 G, and 27 G.
Specifications
Chemical Identifiers
Bioprinting Nozzle
Feature Female Luer Connector
32 mm
Polypropylene
1.26 in.
50 Nozzles
Specifications
Clear
Polypropylene
Feature Female Luer Connector
1.26 in.
50 Nozzles
Bioprinting Nozzle
200 μm
27 G
32 mm
Product Suggestions
Product Suggestions