missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STAMBP Monoclonal Antibody (K56003_18H5), Invitrogen™

Catalog No. MA556128
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556128 200 μL
MA555748 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556128 Supplier Invitrogen™ Supplier No. MA556128
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: KSAVIPEKKD TVKKLKEIAF PKAEELKAEL LKRYTKEYTE YNEEKKKEAE ELARNMAIQQ ELEKEKQRVA QQKQQQLEQE QFHAFEEMIR NQELEKERLK IVQEFGKVDP GLGGPLVPDL EKPSLDVFPT LTVSSIQPSD CHTTVRPAKP PVVDRSLKPG ALSNSESIPT

Cytokine-mediated signal transduction in the JAK-STAT cascade requires the involvement of adaptor molecules. One such signal-transducing adaptor molecule contains an SH3 domain that is required for induction of MYC and cell growth. The protein encoded by this gene binds to the SH3 domain of the signal-transducing adaptor molecule, and plays a critical role in cytokine-mediated signaling for MYC induction and cell cycle progression. Multiple alternatively spliced transcript variants encoding the same protein isoform have been found for this gene.

Specifications

Antigen STAMBP
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K56003_18H5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Stambp
Gene Accession No. O95630
Gene Alias 5330424L14Rik; 5730422L11Rik; AMSH; associated molecule with the SH3 domain of STAM; AW107289; Endosome-associated ubiquitin isopeptidase; MGC126516; MGC126518; MICCAP; mKIAA4198; Stam binding protein; STAM-binding protein; Stambp; testicular secretory protein Li 54
Gene Symbols Stambp
Host Species Mouse
Immunogen Recombinant human STAMBP (immunogen range is 81-250).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10617
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less