missing translation for 'onlineSavingsMsg'
Learn More

Medegen Stainless-Steel Utility Trays

Corrosion resistant and easy to clean

Supplier:  Medegen 30022

 View more versions of this product

Catalog No. 13361D


Only null left

Description

Description

Corrosion resistant and easy to clean

Specifications

Chemical Identifiers

Stainless Steel
Rectangular

Specifications

7.5 L
6.4 cm
Stainless Steel
32 cm
2.5 in.
12.75 in.
20.75 x 12.75 x 2.5 in.
53 cm
Catheter Tray
20.75 in.
Rectangular
7.125 qt.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions