missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Stainless Steel Slim-T™ Flat Head Mop Wringers

These stainless steel, flat head mop wringers are ergonomically crafted from a single piece of stainless steel that easily attaches to the rim of B-7 series buckets. The wringers feature a simple design that leaves mops damp enough to effectively disinfect surfaces, while wringing out excess liquid.

$522.27 - $570.01

Specifications

Material Stainless Steel (304), Electropolished
Autoclavable Autoclavable
Care Instructions Store at Ambient Temperature. Rinse to remove disinfectant before Autoclave or Storage
For Use With (Application) Cleaning Walls, Floors and Ceilings
Quantity 1 Ea.
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Width (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Width (Metric) Price Quantity & Availability  
19035365
View Documents Promotion Details
Micronova™
W92
Dual Side Mop Wringer 40.6 cm
Each for $570.01
Only null left
 
19121802
View Documents Promotion Details
Micronova™
W9
Mop Wringer 45.7 cm
Each for $522.27
Only null left
 
Description

Description

Micronova SlimLine and T-Mop Wringers, Autoclavable: Yes, SlimT Wringer

The Micronova Stainless Steel Slim-T™ Flat Head Mop Wringers work well with the Micronova SlimLine™, SnapMops™, FlatMops™, PadMops™ and PocketMops™ as well as the Slim-T™ and T-Mop series mops. However, the W-9 is preferred with Slim-T and T-Mops, while the W-92, with dual-sided wringing plates, is excellent for PadMops™ and SnapMops™.

  • Lightweight and ergonomic flathead mop wringers
  • Crafted from a single piece of stainless steel with no moving parts
  • Electropolished stainless steel for durability and long life
  • Large lip on W-92 for dual side wringing
  • Designed for the SlimLine™ and T-Mop series as well as other flat head mops
Specifications

Specifications

Stainless Steel (304), Electropolished
Store at Ambient Temperature. Rinse to remove disinfectant before Autoclave or Storage
1 Ea.
Autoclavable
Cleaning Walls, Floors and Ceilings
Non-sterile
Videos
SDS
Documents

Documents