missing translation for 'onlineSavingsMsg'
Learn More

Poltex™ Stainless Steel Pap/Procedure Organizer

Catalog No. 22000770
Encompass
Change view
Click to view available options
Stainless Steel Pap/Procedure Organizer; Material: Stainless Steel (316)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22000770 Stainless Steel Pap/Procedure Organizer; Material: Stainless Steel (316)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22000770 Supplier Poltex™ Supplier No. PAPSMASS

Autoclavable stainless steel organizer for clinical procedures.

  • Your own personal assistant to hold implements and tubes for clinical procedures like cervical cancer screening (pap smear), HPV, Beta Strep, liquid pap, thin prep, etc.
  • Holds culture tubes, swabs, and implements
  • Bottom is enclosed for portability when filled with implements and tubes

Specifications

Product Type Organizer
Color Graphite
For Use With (Application) Holder for B Strep, Thin Prep, Pap Test
For Use With (Equipment) Thin Prep Kit
Width (English) 5.5 in.
Autoclavable Autoclavable
Width (Metric) 140 mm
Material Stainless Steel (316)