missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polar Ware™ Stainless-Steel Instrument Trays
Heavy-duty, seamless trays feature coved corners for easy sterilization
Supplier: Polar Ware™ 1202
Description
Heavy-duty, seamless trays feature coved corners for easy sterilization
- Stainless steel construction
- Available in multiple dimensions
Note:
Covers are not included; sold separately.
Chemical Identifiers
| Stainless Steel | |
| Instrument Tray |
Specifications
| 12.2 x 7.75 x 2.25 in. | |
| 31.1 cm | |
| Instrument Tray | |
| 12.25 in. | |
| Rectangular |
| 5.7 cm | |
| Stainless Steel | |
| 19.7 cm | |
| 2.25 in. | |
| 7.75 in. |