missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Stainless Steel Flask Clamp

OHAUS™ Stainless Steel Flask Clamp is designed to securely grip and position various sizes of flasks.

$180.96 - $267.50

Specifications

Type Flask Clamp
Material Stainless Steel
Color Silver
Jaw Type Four Interlocking
View More Specs

Products 6
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
01922108
View Documents Promotion Details
OHAUS™
30400084
10 mL Flask
Each for $180.96
Only null left
 
01922109
View Documents Promotion Details
OHAUS™
30400085
25 mL Flask
Each for $180.96
Only null left
 
01922110
View Documents Promotion Details
OHAUS™
30400086
50 mL Flask
Each for $189.62
Only null left
 
01922111
View Documents Promotion Details
OHAUS™
30400087
125 mL Flask
Each for $189.62
Only null left
 
01922112
View Documents Promotion Details
OHAUS™
30400088
250 mL Flask
Each for $267.50
Only null left
 
01922113
View Documents Promotion Details
OHAUS™
30400089
500 mL Flask
Each for $211.16
Only null left
 
Description

Description

  • Constructed of stainless steel
  • For use with flasks
  • Designed to securely hold flask in place
Specifications

Specifications

Flask Clamp
Stainless Steel
Silver
Four Interlocking
Videos
SDS
Documents

Documents