missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Stainless Steel Flask Clamp
Description
- Constructed of stainless steel
- For use with flasks
- Designed to securely hold flask in place
Specifications
Specifications
| Type | Flask Clamp |
| Material | Stainless Steel |
| For Use With (Equipment) | 125 mL Flask |
| Description | Flask Clamp, 125 mL |
| Color | Silver |
| Jaw Type | Four Interlocking |