missing translation for 'onlineSavingsMsg'
Learn More

Troemner™ Stainless Steel Electronic Balance Calibration Weights Sets

Catalog No. p-4569646
Change view
Click to view available options
Certifications/Compliance:
NVLAP, ISO9001
Statement of accuracy, ISO9001
Classification:
Class 1
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Classification
02216022 NVLAP, ISO9001 Class 1
022158 Statement of accuracy, ISO9001 Class 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02216022 Supplier Troemner™ Supplier No. 80861110
Only null left

100g to 1kg

  • 2 piece constructions
  • Weights below 1 kg are straight cylinders
  • 1kg weights have groove neare thet top for easy handling
  • Weight set configurations: 1kg, 500g, 300g, 200g, 100g
  • Total capacity of weight set: 2100g
  • UltraClass: With weight tolerances 50% greater than ANSI/ASTM E617 Class 1 tolerances
  • ANSI/ASTM Class 1: Can be used as a reference standard in calibrating other weights and is appropriate for calibrating high precision analytical balances with a readability as low as 0.1mg to 0.01mg
  • ANSI/ASTM Class 2: Appropriate for calibrating high-precision top loading balances with a readability as low as 0.01g to 0.001g
  • ANSI/ASTM Class 3: Appropriate for calibrating balances with moderate precision, with a readability as low as 0.1g to 0.01g
  • ANSI/ASTM Class 4: For calibration of semi-analytical balances and for student use

Order Info

Supplied with impact-resistant polypropylene case.

Specifications

Product Type Electronic Balance Calibration Weight
Certifications/Compliance NVLAP, ISO9001
Classification Class 1
Material Stainless Steel
Weight (Metric) 100 g to 1 kg
Density 7.95g/cm3 at 20°C
Mass 100 g to 1 kg
Includes Cylindrical weight: 1kg, 500g, 300g, 200g, 100g, Plastic case, white Cotton glove
Quantity 5/Set
Packaging Quantity Multiple