Learn More
RETSCH Stainless Steel Distance Sieve For Ultra Centrifugal Mill ZM 200
Catalog No. 08418157
Change view
Click to view available options
Pore Size:
1 mm
Catalog No. Pore Size
08418157 1 mm
1 options
Catalog No. 08418157 Supplier RETSCH Supplier No. 036470259
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Quantity 1Each
Pore Size 1 mm
Product Type Distance Sieve
Material Stainless Steel
For Use With (Equipment) Ultra Centrifugal Mill ZM 200
For Use With (Application) For Temperature-sensitive Materials