Learn More
Mopec Stainless Steel Dissecting Table
Mopec Stainless Steel Dissecting Table can accommodate large or small standard dissection boards
Supplier:  Mopec BC006
 View more versions of this product
Catalog No. BC006
Only null left
Explore available promotions
Description
Description
  • Work surface is tapered toward a centrally located drainage hole
  • 3-inch drain hose (provided) can be connected to a fitting on the underside of the tray
  • Sturdy legs (optional collapsible) can be set on any counter or autopsy table
Specifications
Specifications
Dissecting Table
Standard legs
25 in.
Stainless Steel
Dissecting Boards
18 in.
Product Suggestions
Product Suggestions