missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Stainless Steel Cylindrical Mirrors
Stainless steel cylindrical mirrors are useful for a variety of physical science and optics demonstrations or experiments.
Supplier: Eisco™ PH0529B
Description
- Semi-circular mirrors
- Stainless steel
- Polished surface
- 150 x 75 mm
Specifications
| Stainless Steel | |
| 6.25 in. | |
| Concave Mirror |
| 15.9 cm | |
| Physics and Educational Uses | |
| Half Cylinder |