Learn More
Troemner™ Stainless Steel Class 7 Weight Set
Manufactured from stainless steel. The Troemner™ Stainless Steel Class 7 Weight Set is used for rough weighing operations in physical and chemical laboratories.
Description
- One- and two-piece construction, depending on weight
- Includes certificate of accuracy
- Includes case with latch
- Made in the USA
Specifications
| Weight Set with No Certificate | |
| Stainless Steel | |
| 1.438 in. |
| Class 7 | |
| Plastic | |
| 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg |