Learn More
METTLER TOLEDO™ Stainless Steel Capsule Adapter
Catalog No. 01677031
Change view
Click to view available options
Size:
0
00
000
1
2
3
4
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
01677031 000
01677025 4
01677026 3
01677027 2
01677028 1
01677029 0
01677030 00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01677031 Supplier METTLER TOLEDO™ Supplier No. 30006416
Only null left
Adapter accessories are made with conductive materials to reduce electrostatic charge build up. Our adapters load capsules, vials, bottles, or tubes in precisely the right position and secure them tightly in place for highly accurate dosing.
  • Secures vessel in precise position
  • Compatible with Q3 Sample Changer
  • Conductive materials to reduce electrostatic charge build-up
  • Highly accurate and safe filling
Specifications
For Use With (Equipment) Excellence XPR Automatic Balance
Size 000
Type Stainless Steel Capsule Adapter