Learn More
Brady™ Stainerbondz Super Chemical Resistant Polyester Laboratory Labels
Stainerbondz Super Chemical Resistant Polyester Laboratory Labels (B-481) are designed for use in the slide staining process and other laboratory ID which requires superior chemical resistance.
$278.38 - $318.44
Specifications
Certifications/Compliance RoHS
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Height (English) Price Quantity  
Catalog Number Mfr. No. Quantity Height (English) Price Quantity  
19-140-686
View Documents
Brady™
B33141481
1500/Pk. 0.9 in.
Each for $278.38
Only null left
19-140-693
View Documents
Brady™
B33194481
2500/Pk. 0.75 in.
Each of 2000 for $318.44
Only null left
Description
Description
  • StainerBondz™ B-481 matte white polyester has a permanent acrylic adhesive and is thermal transfer printable with B30 series printers from Brady™
  • B-481 performs well in the commonly used H and E (hematoxilin and eosin) slide staining process
  • Excellent resistance to a wide variety of chemicals, oils and water
  • Service temperature of -112 to 266° F (-80 to 130° C)
Specifications
Specifications
RoHS
None
Matte
Polyester
Laboratory Label
Sample generation, inspection and storage
-112°F to +266°F
40 lb./in.
0.0914 mm
22.86 mm
Acrylic, Permanent
White
Die-cut
Single-side Printable
Non-sterile
-80°C
-80°C to +130°C
0.0033 in.
0.9 in.