missing translation for 'onlineSavingsMsg'
Learn More

Brady™ StainerBondz™ Slide Labels

Made from one of the most solvent-resistant materials in the lab industry

$336.72 - $336.72

Specifications

Certifications/Compliance RoHS 2002/95/EC, MCV 2005/618/EC
Adhesive Type Acrylic, Permanent
Color White
Display Consignment
Finish Matte
View More Specs

Products 2
Catalog Number Mfr. No. Labels per Row Price Quantity  
Catalog Number Mfr. No. Labels per Row Price Quantity  
22-500-558
missing translation for 'viewDocuments'
Brady™
THT16448125SC
1
Pack of 2500 Each for $336.72
Only null left
 
22-500-561
missing translation for 'viewDocuments'
Brady™
THT20748125SC
4
Pack of 2500 Each for $336.72
Only null left
 
Description

Description

  • Made of B-481 material and coupled with R6400 series ribbon
  • White matte polyester
  • Remains resistant when exposed to xylene and other chemicals commonly used in the slide staining process
  • Bar codes remain readable and labels stay adhered to the slide
  • No print removal, no label buckling, and no discoloration
  • Brady IP™ enabled

Applications:

Slide Label Identification

Specifications

Specifications

RoHS 2002/95/EC, MCV 2005/618/EC
White
Matte
Polyester
Thermal Transfer
Non-sterile
BBP™ 11, Brady 1244, Brady 1344 Printer
0.9 in.
2.28 cm
2.28 cm
2.54 cm
Acrylic, Permanent
Consignment
Label
Single-side Printable
Label
General identification, Slide label ID
2500/Roll
0.9 in.
0.9 in.
1 in.
Videos
SDS
Documents

Documents