missing translation for 'onlineSavingsMsg'
Learn More

Stachyose Hydrate 98.0+%, TCI America™

Catalog No. S03971G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03971G 1 g
S03975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03971G Supplier TCI America Supplier No. S03971G

Chemical Identifiers

CAS 54261-98-2
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.58
MDL Number MFCD00149457
InChI Key UQZIYBXSHAGNOE-CYDMKDQUNA-N
Synonym beta-D-Fructofuranosyl O-alpha-D-Galactopyranosyl-(1-6)-O-alpha-D-galactopyranosyl-(1-6)-alpha-D-glucopyranoside, Lupeose, Manneotetrose
PubChem CID 131801001
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-({[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-({[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}methyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 110°C
Color White
Quantity 1 g
Formula Weight 666.58
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Stachyose Hydrate