missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Stachyose hydrate, 97+%

Catalog No. p-6827168
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226085000 500 mg Glass bottle
AC226080010 1 g Glass bottle
AC226080050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC226085000 Supplier Thermo Scientific Chemicals Supplier No. 226085000

CAS: 54261-98-2 | C24H42O21 | 666.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54261-98-2
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.58
MDL Number MFCD00149457
InChI Key UQZIYBXSHAGNOE-CYDMKDQUNA-N
PubChem CID 131801001
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Color Brown to Colorless
Physical Form Liquid
Melting Point 95.0°C to 105.0°C
Quantity 500 mg
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97% min. (ELSD) (HPLC)
Specific Rotation +129
Solubility Information 5% in water, Clear to slightly hazy, Colorless
IUPAC Name 2-[[6-[[6-[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;trihydrate
Formula Weight 666.58
Percent Purity ≥97%
Chemical Name or Material Stachyose hydrate, 98.5%
Show More Show Less

RUO – Research Use Only