missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ST3GAL1 293T Cell Transient Overexpression Lysate(Denatured)

Catalog No. 89339488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89339488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89339488 Supplier Abnova™ Supplier No. H00006482T02

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Gene Accession No. NM_003033.2
Plasmid pCMV-ST3GAL1 full-length
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.