missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SSTR3 Recombinant Rabbit Monoclonal Antibody (JE63-87)
GREENER_CHOICE

Catalog No. PIMA544784
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544784 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA544784 Supplier Invitrogen™ Supplier No. MA544784
Only null left

Rabbit Recombinant Monoclonal Antibody

SSTR3 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

Expression of Somatostatin Receptor Type 3 (SSTR3) has been reported in brain, breast, eye, gastrointestinal tract, pancreas, and thyroid. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.

Specifications

Antigen SSTR3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE63-87
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Sstr3
Gene Accession No. P32745
Gene Alias Smstr28; Smstr3; Smstr-3; somatostatin receptor; somatostatin receptor 28; somatostatin receptor 3; somatostatin receptor subtype 3; somatostatin receptor type 3; SS3R; SS-3-R; SS3-R; SSR-28; sst3; SSTR; SSTR3
Gene Symbols Sstr3
Host Species Rabbit
Immunogen Synthetic peptide within human SSTR3 aa 369-418/418.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6753
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less